Sleeves for ground joints
Lieferant: GLASWARENFABRIK KARL HECHT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
42388010 42388019 42388024 42388029
HECH42388010EA 20 EUR InStock
HECH42388010 HECH42388019 HECH42388024 HECH42388029
Sleeves for ground joints
Normschliff-Manschetten und Faltenbälge Schliffmanschetten
Disposable sleeves for ground joints and stoppers, prevent the ground joints from sticking together.
  • Made of PTFE
  • Self-lubricating
  • Outstanding chemical resistance at temperatures
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge