Stoppers for standard ground joints, octogonal, HDPE
Lieferant: GLASWARENFABRIK KARL HECHT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
42990007 42990010 42990012 42990014 42990019 42990024 42990029 42990034 42990045
HECH42990007EA 0.5 EUR InStock
HECH42990007 HECH42990010 HECH42990012 HECH42990014 HECH42990019 HECH42990024 HECH42990029 HECH42990034 HECH42990045
Stoppers for standard ground joints, octogonal, HDPE
Stopfen PE/LDPE/HDPE-Stopfen
High-density polyethylene.
  • Acid and alkali resistant
  • Octogonal
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge