Thermostatic water bath
Lieferant: GLASWARENFABRIK KARL HECHT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
43180010
HECH43180010EA 1490 EUR InStock
HECH43180010
Thermostatic water bath
Bäder Heizbäder
The Thermostatic waterbath, CE certified is fully electronic wear-free temperature control with digital display. Temperature control with adjustable nominal value limit and alarm circuit.
  • Themperature stability of ± 0,1 K
  • Operating temperature range with external cooling of −30 °C upto +100 °C
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge