Weather station
Lieferant: GLASWARENFABRIK KARL HECHT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
43655010
HECH43655010EA 58 EUR InStock
HECH43655010
Weather station
Wetterstationen
This wireless weather station with sensor unit combines the features of a barometer (tendency), thermometer and hygrometer. The sensor unit can be placed at a distance of max. 100 m from the main station.
  • Radio controlled clock
  • With LCD display and LED indicator
  • Low battery warning
  • Stand-up device, wall mounting is possible
  • Range dependent on building structure and sensor unit placement
W×D×H (Station): approx. 102×33×180 mm / 230 g
W×D×H (Sensor unit): approx. 43×26×160 mm / 75 g
Lieferumfang: Supplied with 3 batteries AA 1,5 V (LR 6).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge