Draining racks
Lieferant: GLASWARENFABRIK KARL HECHT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
49027010 49028010
HECH49027010EA 98 EUR InStock
HECH49027010 HECH49028010
Draining racks
Gestelle Abtropfgestelle
Made of wire, plastic coated.
  • Good chemical resistance
  • Good temperature resistance
  • Easy to clean
Suitable for hanging or free standing test tubes and flasks with a plastic trough.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge