Hei-CHILL 5000 Circulating Chillers
Lieferant: Heidolph Instruments GmbH & Co.KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
591-01710-00
HEID591-01710-00EA 0 EUR InStock
HEID591-01710-00
Hei-CHILL 5000 Circulating Chillers
Umwälzthermostate Geschlossene Umwälzsysteme Kälteumwälzthermostate
Comprehensive spectrum of services for demanding temperature control tasks with a cooling capacity of 5000 W.
  • Filling opening at the top, drain tap at the rear
  • Integrated programmer with 150 segments, can be divided into 5 programs
  • Electronic level indicator and low-level alarm
  • SmartCool system for digital, energy-saving cooling control, including automatic compressor control
  • Clearly arranged control elements, large TFT display and membrane keypad
  • USB interface and an alarm contact are integrated as standard features
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge