Base Cart, Hei-CHILL 5000
Lieferant: Heidolph Instruments GmbH & Co.KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
591-01720-00
HEID591-01720-00EA 0 EUR InStock
HEID591-01720-00
Base Cart, Hei-CHILL 5000
Umwälzthermostate Geschlossene Umwälzsysteme Kälteumwälzthermostate
The Hei-CHILL 5000 Base cart is the most compact and strongest undercounter chiller with a cooling capacity of 5000 W.
  • Optimally designed as space-saving solution to be used with a large-scale evaporator and other peripheral devices, this compact undercounter chiller can be perfectly placed among one of our Hei-VAP Industrial models
  • The power supply of the rotary evaporator is ensured by a power socket at the rear of the chiller
  • Despite its small footprint, this chiller also provides space for a vacuum pump and mounting possibilities for the automatic module Hei-VOLUME Distimatic Pro
  • With its cooling capacity of up to 5 kW, this recirculation chiller is extremely powerful
  • Its large reservoir is particularly useful for peak loads, and a high flow rate
  • Suitable even for demanding liquids like low-boilers and continuous evaporation, combined with the automatic module Hei-VOLUME Distimatic Pro
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge