Colour stripe silicone tubing, HelixMark®
Lieferant: HELIX MEDICAL EUROPE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
61-079-21 61-079-27 61-080-21 61-080-23 61-080-26 61-080-27 61-080-30 61-080-32 61-079-23 61-079-26 61-079-30 61-079-32
HELI61-079-21EA 0 EUR InStock
HELI61-079-21 HELI61-079-27 HELI61-080-21 HELI61-080-23 HELI61-080-26 HELI61-080-27 HELI61-080-30 HELI61-080-32 HELI61-079-23 HELI61-079-26 HELI61-079-30 HELI61-079-32
Colour stripe silicone tubing, HelixMark®
Schläuche Transferschlauch
Distinctive silicone colour striping enables users to recognise tube lines at a glance by application, tubing size, or functional area.
  • Fully autoclavable and sterilisable
  • Ideal for use in transfer operations as well as other applications requiring non-leachable material
  • Material certification in every package
  • Lot traceability
Designed to suit a wide array of laboratory applications or pharmaceutical processes. All tubes provide exceptional biocompatibility, having undergone studies according to ISO 10993-1 and meeting USP Class VI requirements. The production process ensures that the silicone stripe is integrated into the tube wall without touching or impeding the interior passage.
Zertifizierungen: Tubing is manufactured under strict good manufacturing practices (GMP) in a FDA registered medical device facility.
Bestellinformation: Tubing is packaged in 15,24 m coils and double-bagged in crush-resistant boxes. Material certification and lot traceability is included in every box.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge