Racks for chest freezers
Lieferant: HERAEUS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
398187 398185 398186 398184
HERA398187EA 132 EUR InStock
HERA398187 HERA398185 HERA398186 HERA398184
Racks for chest freezers
Gestelle Kryogestelle
Use Thermo Scientific™ Chest freezer racks with TDE Series ultra-low temperature chest freezers as well as legacy Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-low temperature chest freezers.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge