Gestelle für Revco Tieftemperatur-Gefrierschränke
Lieferant: HERAEUS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
398331 398324 398332 398330 398329 398328 398327 398325 398326
HERA398331EA 222 EUR InStock
HERA398331 HERA398324 HERA398332 HERA398330 HERA398329 HERA398328 HERA398327 HERA398325 HERA398326
Gestelle für Revco Tieftemperatur-Gefrierschränke
Gestelle Kryogestelle
Maximise the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for Revco™ UxF and HERAfreeze™ HFU T freezers.
Bestellinformation: Warranty varies by country, please contact us for details.
Lieferumfang: Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge