HAEMAFlex™ 6/8 Swinging Bucket Rotors
Lieferant: HERAEUS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
75003881 75003861
HERA75003881EA 18700 EUR InStock
HERA75003881 HERA75003861
HAEMAFlex™ 6/8 Swinging Bucket Rotors
Zentrifugen Rotoren
Address lab’s regular- to medium-throughput blood processing needs with the Thermo Scientific™ HAEMAFlex™ six swinging bucket rotor with 6×550 ml blood bag capacity or, utilising the same bucket system, upgrade to 8×550 ml bags with the Thermo Scientific™ HAEMAFlex™ eight swinging bucket rotor.
  • Eco-Spin™ technology provides energy savings up to 64% based on an engineering evaluation of windshielded and non-windshielded designs of the same rotor body
  • Stainless steel rotor body, aluminum windshield and bucket, polypropylene bottle
High-throughput sample processing with the flexibility to match your current and future needs with 6 or 8×550 ml blood bag (6 or 8×1000 ml) capacity, both available with the same bucket selections. Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.
Compatible with Thermo Scientific™ Auto-Lid™ and Auto-ID™ functions enabling instant rotor lid opening, storage and rotor identification to save time and protects the integrity of your samples.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge