Ultra low temperature freezers, with 5 inner doors, –86 °C, TDE series
Lieferant: HERAEUS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
TDE30086FV5ID TDE40086FV5ID TDE50086FV5ID TDE60086FV5ID TDE30086FV5I-U TDE40086FV5I-U TDE50086FV5I-U TDE60086FV5I-U
HERATDE30086FV5IDEA 14100 EUR InStock
HERATDE30086FV5ID HERATDE40086FV5ID HERATDE50086FV5ID HERATDE60086FV5ID HERATDE30086FV5I-U HERATDE40086FV5I-U HERATDE50086FV5I-U HERATDE60086FV5I-U
Ultra low temperature freezers, with 5 inner doors, –86 °C, TDE series
Gefriergeräte Ultratiefkühlgeräte
Thermo Scientific™ TDE Series ultra-low temperature freezers, powered by H-drive, feature four upright models with 5 Inner doors, maximizing storage capacity from 30000 up to 60000 2 ml vials. The TDE Series is a sustainable and reliable solution with ultimate sample security and operational savings for every lab. Sustainability features include natural refrigerants and water-blown foam, compliant with many sustainability standards.
  • Powered by H-drive, save up to 25% in energy usage and heat rejection over conventional refrigerant ultra-low freezers
  • Easy-to-use set-up wizard, event log, temperature mapping and data downloads
  • Capacitive touch even when using latex gloves
  • Touch-screen datalogging and temperature mapping eliminates the need for a separate chart recorder
  • Built-in USB port for exporting data
  • Advanced security features: Alarm settings, password-protected controller, padlock-compatible doors and optional access card entry
  • Ergonomic design: Comfortable door handle, eye-level touch-screen and shelf-level placement that does not require floor-level loading of racks
  • 5 year warranty
The units have five inner doors which help minimize ambient temperature changes during sample retrieval. Low peak variation helps ensure sample protection throughout entire cabinet, they have fast door opening recovery times, as little as 11 minutes and the low noise level, below 52 dBA, allows these quiet freezers to be brought out of the hallway and into the laboratory.
Bestellinformation: Accessories must be ordered separately. Field installed options and accessories must be installed by a qualified professional.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge