Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Ultra low temperature freezers, with 4 inner doors, –40 °C, TDE series

Lieferant: HERAEUS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

TDE40040LV TDE40040FV TDE50040FV TDE60040FV
HERATDE40040LVEA 12600 EUR InStock
HERATDE40040LV HERATDE40040FV HERATDE50040FV HERATDE60040FV
Ultra low temperature freezers, with 4 inner doors, –40 °C, TDE series
Gefriergeräte Ultratiefkühlgeräte
Thermo Scientific™ TDE Series ‒40 °C ultra-low temperature freezers, powered by H-drive, feature three upright models, maximizing storage capacity from 40000 up to 60000 2 ml vials. The TDE Series is a sustainable and reliable solution with ultimate sample security and operational savings for every laboratory.

  • Powered by H-drive, save up to 25% in energy usage and heat rejection over conventional refrigerant ultra-low freezers
  • Easy-to-use set-up wizard, event log, temperature mapping and data downloads
  • Capacitive touch even when using latex gloves
  • Touch-screen datalogging and temperature mapping eliminates the need for a separate chart recorder
  • Built-in USB port for exporting data
  • Advanced security features: Alarm settings, password-protected controller, padlock-compatible doors and optional access card entry
  • Ergonomic design: Comfortable door handle, eye-level touch-screen and shelf-level placement that does not require floor-level loading of racks
  • 5 year warranty
  • Stainless steel interior

The units have four inner doors which help minimize ambient temperature changes during sample retrieval. Low peak variation helps ensure sample protection throughout entire cabinet. The low noise level, below 52 dBA, allows these quiet freezers to be brought out of the hallway and into the laboratory.

Style: Upright
Amperage: 3,2/10
Plug type: EU

Monitoring options: Alarm, recorder
Setpoint security: Yes
Connectivity: Remote alarm connections, 4 to 20 mA, RS485, and dry contacts
Smart view compatibility: Smart-Vue compatible

Bestellinformation: Accessories must be ordered separately. Field installed options and accessories must be installed by a qualified professional.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge