Valve supplies for HPLC system
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0100-1849 0100-1850 0100-1851 0100-1853 0101-0921 0100-2195 0100-2088 0100-2089 0101-1050 0101-1257 0101-1268 0100-2231 0101-1385 0101-1390 0101-1409 0101-1417 0101-1416 0101-1422 5068-0007 5067-4117 5068-0006
HEWL0100-1849EA 0 EUR InStock
HEWL0100-1849 HEWL0100-1850 HEWL0100-1851 HEWL0100-1853 HEWL0101-0921 HEWL0100-2195 HEWL0100-2088 HEWL0100-2089 HEWL0101-1050 HEWL0101-1257 HEWL0101-1268 HEWL0100-2231 HEWL0101-1385 HEWL0101-1390 HEWL0101-1409 HEWL0101-1417 HEWL0101-1416 HEWL0101-1422 HEWL5068-0007 HEWL5067-4117 HEWL5068-0006
Valve supplies for HPLC system
Chromatographie-Zubehör Chromatographie-Ersatzteile
Vespel is a polyimide with low wear and high chemical resistance. Vespel tolerates a pH range of 0 to 10.
PEEK offers a high chemical resistance and flexibility and will tolerate the entire pH range from 0 to 14.
Tefzel is ideal for use in applications where PEEK cannot be used, such as methylene chloride or DMSO in higher concentrations.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge