DB-TPH GC columns, J&W
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
123-1632
HEWL123-1632EA 903 EUR InStock
HEWL123-1632
DB-TPH GC columns, J&W
Chromatographiesäulen
DB-TPH is specifically designed for the analysis of total petroleum hydrocarbons (TPH), soil analysis, and leaking underground fuel tanks (LUFT).
  • Specifically designed for the analysis of total petroleum hydrocarbons (TPHs), soil analysis, and LUFT
  • Three analyses in one injection – gas range organics, diesel range organics and motor oil
  • Fast run time
  • Bonded and cross-linked
  • Solvent rinsable
DB-TPH columns feature three analyses in a single injection (gas range organics, diesel range organics, and motor oil) and are fast run time columns. With solvent rinsable functionality and an operating temperature range of −10 to +320 °C, these high quality and extensively QC tested columns are excellent for petroleum analysis applications.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge