GC columns for blood alcohol concentration (BAC)
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
123-9334UI 123-9434UI 125-9434UI 125-9334UI
HEWL123-9334UIEA 882 EUR InStock
HEWL123-9334UI HEWL123-9434UI HEWL125-9434UI HEWL125-9334UI
GC columns for blood alcohol concentration (BAC)
Chromatographiesäulen
These columns are suitable for the analysis of Blood Alcohol Concentration (BAC) using Headspace GC dual FID or headspace GC/MS.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge