Sample Loops for Rheodyne® Valves
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0100-1921 0100-1922 0100-1923 0100-1924 0101-0376 0101-0377 0101-0378 0101-0379 1535-4860 0101-1219 0101-1226 0101-1227 0101-1228 0101-1229 0101-1230 0101-1234 0101-1235 0101-1236 0101-1237 0101-1238 0101-1239 0101-1240 0101-1241 0101-1242 0101-1243 0101-1244 0101-1245 0101-1246 0101-1247 0101-1248 0101-1249 0101-1250 0101-1251
HEWL0100-1921EA 0 EUR InStock
HEWL0100-1921 HEWL0100-1922 HEWL0100-1923 HEWL0100-1924 HEWL0101-0376 HEWL0101-0377 HEWL0101-0378 HEWL0101-0379 HEWL1535-4860 HEWL0101-1219 HEWL0101-1226 HEWL0101-1227 HEWL0101-1228 HEWL0101-1229 HEWL0101-1230 HEWL0101-1234 HEWL0101-1235 HEWL0101-1236 HEWL0101-1237 HEWL0101-1238 HEWL0101-1239 HEWL0101-1240 HEWL0101-1241 HEWL0101-1242 HEWL0101-1243 HEWL0101-1244 HEWL0101-1245 HEWL0101-1246 HEWL0101-1247 HEWL0101-1248 HEWL0101-1249 HEWL0101-1250 HEWL0101-1251
Sample Loops for Rheodyne® Valves
Chromatographie-Zubehör Chromatographie-Probenschläuche
These PEEK sample loops are inert to most organic solvents. The stainless-steel sample loops are not interchangeable due to the change in port angle.
  • Stainless steel loops are square cut and free of burrs for a flush connection
  • Flexible PEEK loops have a clean, straight cut for low dead volume connections
Accuracy of large PEEK loops is ±14%, intermediate loops ±21%, small loops ±65%. Accuracy of large metal loops is ±5%, intermediate loops ±10%, small loops ±30%
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge