Flame ionisation detector (FID) supplies
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
18710-20119 18710-60170 19231-20910 19231-20940 19231-20970 19231-21050 19231-21060 19231-60680 19231-80520 19231-80530 19244-80560 19244-80610 19301-00150 19301-60660 5080-4978 5180-4150 5180-4152 5180-4153 5180-4165 5181-3311 5182-3450 8710-1346 8710-1561 9301-0985 G1531-20690 G1531-20700 G1531-60680 G1531-60690 G1531-80620 G1531-21090 5188-5372 G3431-60680
HEWL18710-20119EA 0 EUR InStock
HEWL18710-20119 HEWL18710-60170 HEWL19231-20910 HEWL19231-20940 HEWL19231-20970 HEWL19231-21050 HEWL19231-21060 HEWL19231-60680 HEWL19231-80520 HEWL19231-80530 HEWL19244-80560 HEWL19244-80610 HEWL19301-00150 HEWL19301-60660 HEWL5080-4978 HEWL5180-4150 HEWL5180-4152 HEWL5180-4153 HEWL5180-4165 HEWL5181-3311 HEWL5182-3450 HEWL8710-1346 HEWL8710-1561 HEWL9301-0985 HEWLG1531-20690 HEWLG1531-20700 HEWLG1531-60680 HEWLG1531-60690 HEWLG1531-80620 HEWLG1531-21090 HEWL5188-5372 HEWLG3431-60680
Flame ionisation detector (FID) supplies
Chromatographie-Zubehör Chromatographie-Ersatzteile
Before ordering parts for FID maintenance, determine which type of FID is installed on your GC. The FID is available in two versions: Dedicated, capillary optimized: for capillary columns only. Adaptable: for packed or capillary columns.
To determine the type of FID installed on your GC, open the oven door and examine the fitting at the base of the detector.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge