GC columns, J&W HP-35
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
19091G-113 19091G-131 19091G-133 19091G-213
HEWL19091G-113EA 795 EUR InStock
HEWL19091G-113 HEWL19091G-131 HEWL19091G-133 HEWL19091G-213
GC columns, J&W HP-35
Chromatographiesäulen
HP-35 is a mid-polarity column, inert to active solutes and ideal for conformational analyses.
  • (35%-Phenyl)-methylpolysiloxane
  • Mid polarity-slightly less polar than DB-35
  • Inert to active solutes
  • Ideal for confirmational analyses
  • Bonded and cross-linked
This phase has been specified by EPA method 8081 and U.S.P G-42. The intermediate polarity of HP-35 makes it a good alternative for the analysis of pesticides, herbicides, pharmaceuticals, and amines.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge