Syringe filters, Econofilters
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5190-5268 5190-5264 5190-5265 5190-5266 5190-5267 5190-5310 5190-5280
HEWL5190-5268EA 760 EUR InStock
HEWL5190-5268 HEWL5190-5264 HEWL5190-5265 HEWL5190-5266 HEWL5190-5267 HEWL5190-5310 HEWL5190-5280
Syringe filters, Econofilters
Filter Spritzenvorsatzfilter
These syringe filters have a PP housing.
  • Fast and efficient filtration
  • Low level of extractables
  • High flow rate
  • Maximum sample loading capacity
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge