5982-1182
5982-5750
5982-5751
5982-5753
5982-1082
5982-1382
5982-1882
5982-2082
5982-2282
5982-4382
5982-4482
5982-5752
5982-8082
5982-8382
HEWL5982-1182EA
0
EUR
InStock
HEWL5982-1182
HEWL5982-5750
HEWL5982-5751
HEWL5982-5753
HEWL5982-1082
HEWL5982-1382
HEWL5982-1882
HEWL5982-2082
HEWL5982-2282
HEWL5982-4382
HEWL5982-4482
HEWL5982-5752
HEWL5982-8082
HEWL5982-8382
Bulk sorbents, SampliQ
Chromatographie-Sorbentien
Flash-Chromatographie-Sorbentien
QuEChERS Bulk Salts and Sorbents make sample prep easy.
- Prepackaged QuEChERS Kits are an easy way to capture the time-saving benefits of QuEChERS sample preparation
- Extraction kits with preweighed anhydrous salts in sealed packets allow you to add salts after you add organic solvent to your sample – avoiding an exothermic reaction that can compromise analyte recovery
- Dispersive kits with sorbents and salts supplied in 2 or 15 ml centrifuge tubes accommodate the aliquot volumes specified by current AOAC and EN methodologies
- Universal dispersive kits provide excellent recoveries and reproducibility for all types of fruits and vegetables
- Ceramic homogenizers break up salt agglomerates, promoting consistent sample extraction and increasing product recovery during extraction and dispersion; shaking time reduced from 60 to 20 seconds
If you prefer to pack your own tubes for QuEChERS, these bulk salts and sorbents provide high-quality materials for that purpose.