Bulk sorbents, SampliQ
Lieferant: AGILENT HEWLETT PACKARD
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5982-1182 5982-5750 5982-5751 5982-5753 5982-1082 5982-1382 5982-1882 5982-2082 5982-2282 5982-4382 5982-4482 5982-5752 5982-8082 5982-8382
HEWL5982-1182EA 0 EUR InStock
HEWL5982-1182 HEWL5982-5750 HEWL5982-5751 HEWL5982-5753 HEWL5982-1082 HEWL5982-1382 HEWL5982-1882 HEWL5982-2082 HEWL5982-2282 HEWL5982-4382 HEWL5982-4482 HEWL5982-5752 HEWL5982-8082 HEWL5982-8382
Bulk sorbents, SampliQ
Chromatographie-Sorbentien Flash-Chromatographie-Sorbentien
QuEChERS Bulk Salts and Sorbents make sample prep easy.
  • Prepackaged QuEChERS Kits are an easy way to capture the time-saving benefits of QuEChERS sample preparation
  • Extraction kits with preweighed anhydrous salts in sealed packets allow you to add salts after you add organic solvent to your sample – avoiding an exothermic reaction that can compromise analyte recovery
  • Dispersive kits with sorbents and salts supplied in 2 or 15 ml centrifuge tubes accommodate the aliquot volumes specified by current AOAC and EN methodologies
  • Universal dispersive kits provide excellent recoveries and reproducibility for all types of fruits and vegetables
  • Ceramic homogenizers break up salt agglomerates, promoting consistent sample extraction and increasing product recovery during extraction and dispersion; shaking time reduced from 60 to 20 seconds
If you prefer to pack your own tubes for QuEChERS, these bulk salts and sorbents provide high-quality materials for that purpose.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge