Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Screw top vials and caps, 2 ml

Lieferant: Agilent

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

5183-2070 5183-2072 5067-0237 8010-0426
HEWL5183-2070EA 52 EUR InStock
HEWL5183-2070 HEWL5183-2072 HEWL5067-0237 HEWL8010-0426
Screw top vials and caps, 2 ml
Vials
Vials with 2 ml wide opening are constructed from type 33 and type 51 COE borosilicate glass, with low metal content, to protect your sample from destabilizing or leaching.

  • Uniformly flat bottom ensures compatibility with inserts
  • Precision-formed neck for optimal robotic arm handling
  • Unique thread design for consistently secure seal
  • Optional ceramic write-on spot with fill marks
  • Rigorous quality assurance for dimensional consistency from lot to lot

Wide opening vials have a 40% larger opening than standard opening vials.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge