Gel filtration chromatography columns, ZORBAX
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
820675-111 820950-908 820950-911 884973-701 884973-901 884973-902 820212-911 877974-901 877974-910
HEWL820675-111EA 0 EUR InStock
HEWL820675-111 HEWL820950-908 HEWL820950-911 HEWL884973-701 HEWL884973-901 HEWL884973-902 HEWL820212-911 HEWL877974-901 HEWL877974-910
Gel filtration chromatography columns, ZORBAX
Chromatographiesäulen
These ZORBAX GF-250 and GF-450 size exclusion (gel filtration) columns are ideal for the size separations of proteins and other biomolecules.
  • High efficiency and reproducibility with short analysis time
  • Hydrophilic diol bonded phase for good protein recovery
  • Compatible with organic modifiers and denaturants
  • Wide usable pH range (pH 3 to 8)
Some common applications includes separations of protein monomers, dimers and aggregates, desalting, protein molecular weight estimation and separations of modified proteins.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge