GF-250/450 columns, ZORBAX®
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
884973-761
HEWL884973-761EA 0 EUR InStock
HEWL884973-761
GF-250/450 columns, ZORBAX®
Chromatographiesäulen
ZORBAX® GF-250 and GF-450 size exclusion (gel filtration) columns are ideal for size separations of proteins and other biomolecules.
  • High efficiency and reproducibility with short analysis time
  • Semi-prep and prep column dimensions
  • Compatible with organic modifiers and denaturants
  • Wide usable pH range (3 to 8)
The separation range is 4000 to 900000 for globular proteins when using GF-250 and GF-450 columns in series. The GF-250/GF-450 size exclusion columns have a hydrophilic diol bonded phase for high recovery of proteins (typically greater than 90%) and a unique zirconia modification of the silica for a pH operating range from 3 to 8. The GF-250 and GF-450 columns are packed with precisely sized porous silica microspheres with narrow pore size and particle size distributions. The result is a highly efficient, rugged and reproducible size exclusion column that can be used for both analytical and preparative separations of proteins with flow rates of up to 3 ml/min.
These columns are compatible with organic modifiers ( less than 25%) and denaturants in the mobile phase to reduce protein aggregation. Some common applications include separations of protein monomers, dimers and aggregates, desalting, protein molecular weight estimation and separations of modified proteins. View our full range of bioHPLC columns for your area of interest, get resources, and take advantage of special offers.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge