Conductivity/TDS meter, bench, 3200C
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
G4384A
HEWLG4384AEA 0 EUR InStock
HEWLG4384A
Conductivity/TDS meter, bench, 3200C
Elektrochemische Messgeräte Leitfähigkeitsmessgeräte
This full-range meter measures conductivity, resistivity, total dissolved solid (TDS), salinity, and present solution temperature. Ideal for use in environmental, petrochemical, biological and food applications.
  • Provides quick and precise measurements under any conditions
  • Temperature compensation
Bestellinformation: Supplied without electrode which needs to be ordered separately.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge