Nitrogen and Sulphur chemiluminescence detectors supplies
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
G1530-80650 G6600-60037 G6600-60038 G6600-67007 G6600-67008 G6600-80042 G6600-80043 G6600-80044 G6600-80045 G6600-80050 G6600-80051 G6600-80063 G6600-85000 G6600-85001 G6600-85002 G2630-60710 G2933-85001 G6600-60035 G6600-80018
HEWLG1530-80650EA 398 EUR InStock
HEWLG1530-80650 HEWLG6600-60037 HEWLG6600-60038 HEWLG6600-67007 HEWLG6600-67008 HEWLG6600-80042 HEWLG6600-80043 HEWLG6600-80044 HEWLG6600-80045 HEWLG6600-80050 HEWLG6600-80051 HEWLG6600-80063 HEWLG6600-85000 HEWLG6600-85001 HEWLG6600-85002 HEWLG2630-60710 HEWLG2933-85001 HEWLG6600-60035 HEWLG6600-80018
Nitrogen and Sulphur chemiluminescence detectors supplies
Chromatographie-Zubehör Chromatographie-Ersatzteile
Nitrogen Chemiluminescence Detector (NCD) is a nitrogen-specific detector that produces a linear and equimolar response to nitrogen compounds based on a chemiluminescent reaction of NO with ozone.
Sulphur Chemiluminescence Detector (SCD) for the analysis of sulphur compounds.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge