Moisture traps, MT series
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MT120-2 MT120-4 MT200-2 MT200-4 MT400-2
HEWLMT120-2EA 216 EUR InStock
HEWLMT120-2 HEWLMT120-4 HEWLMT200-2 HEWLMT200-4 HEWLMT400-2
Moisture traps, MT series
Filter Gasfilter
Indicating moisture traps are available in plastic and glass bodies.
  • Glass body traps are used when potential contaminants from plastic trap bodies are a concern
  • Glass traps are normally encased in a protective, plastic shrink wrap or a high impact plastic shield (outer trap body)
Glass and plastic bodied traps are usually pressure tested at 10342 bar, thus they are safe for use at the typical pressures required by the GC.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge