Avantor® Hichrom PEEKsil™, Tubing Sleeves, PEEK, MicroTight
Lieferant: Hichrom Limited
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
HICHHI-171EA 46.9 EUR InStock
HICHHI-171
Avantor® Hichrom PEEKsil™, Tubing Sleeves, PEEK, MicroTight
Schläuche Chromatographie-Schläuche
PEEK.
  • Colour-coded for easy Int.Ø identification
  • Temperature stable up to 125 °C
These tubing sleeves are used with MicroTight fittings to connect capillary tubing. The Int.Ø of the sleeve should be 0,001 to 0,002" larger than the Ext.Ø of the capillary tubing.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge