Avantor® Hichrom, Static Mixing Tee
Lieferant: Hichrom Limited
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
HICHHI-454EA 227 EUR InStock
HICHHI-454
Avantor® Hichrom, Static Mixing Tee
Fittings und Verbinder
These static mixing tee are ideal for microbore or analytical gradient HPLC. It has a low swept volume (2,2 µl) and is designed for flow rates of 0,5 to 3 ml/min.
  • PEEK body with 2-piece finger tight fittings
  • Low swept volume
  • Pressure rated to 345 bar
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge