Columns for large biomolecule analysis, CIMmultus™
Lieferant: HICHROM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
311.6157-6 311.3010-1.3 311.5114-1.3 311.5113-2 311.5113-1.3 311.5113-6 311.8140-2 311.6157-2 346.0001-2 411.8136-2 411.5114-1.3 411.5113-2 411.8140-2 411.8140-1.3 411.6157-2
HISM311.6157-6EA 0 EUR InStock
HISM311.6157-6 HISM311.3010-1.3 HISM311.5114-1.3 HISM311.5113-2 HISM311.5113-1.3 HISM311.5113-6 HISM311.8140-2 HISM311.6157-2 HISM346.0001-2 HISM411.8136-2 HISM411.5114-1.3 HISM411.5113-2 HISM411.8140-2 HISM411.8140-1.3 HISM411.6157-2
Columns for large biomolecule analysis, CIMmultus™
Chromatographiesäulen
CIMmultus™ columns are based on an innovative monolithic technology. Distinguished by large pores/flow through channels these columns are designed for the production, purification, and analytics of large proteins (IgG, IgM, PEGylated proteins), viruses (AAV, adenovirus, lentivirus, influenza, bacteriophages), VLPs, exosomes and pDNA for example.
  • Rapid, high resolution analysis
  • Flow independent separation
  • High sensitivity and reproducibility
  • Operation at high volumetric flow rates
  • Larger scale columns available and in cGMP compliant formats
Distinguished by their flow independent resolution, these columns offer high efficiency, high speed, and high yield in downstream processing for large biomolecules. Due to their performance properties, CIMmultus™ columns can be used as single-use disposable or multi-use columns.
Zertifizierungen: USP Class VI.
Achtung! Bitte beachten: Do not expose the column housing to pure acetone.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge