Embedding system, Technovit® 3040
Lieferant: HERAEUS KULZER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
64708805
HKUL64708805EA 168 EUR InStock
HKUL64708805
Embedding system, Technovit® 3040
Gewebeeinbetter
Technovit 3040 is a quickly hardening two-component plastic with a methyl meth acrylate base (MMA).
  • Chemical composition allows for a stable permanent bond with the hardened block
  • Hardens and the Histobloc tightly bonds to the specimen
  • Specimen can now be removed from the form and is ready to be cut. Through blocking with the very hard
Used to block or fasten embedded plastic specimens via a block or emmbedding specimen.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge