Uranium ICP and ICP-MS Standards, (CRMs), Fluka™
Lieferant: HONEYWELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
E1361-100ML E1430-100ML
HONCE1361-100MLEA 135.51 EUR InStock
HONCE1361-100ML HONCE1430-100ML
Uranium ICP and ICP-MS Standards, (CRMs), Fluka™
Standards Kationischer Standards Uran-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Zertifizierungen: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge