Combined Seven Anion Standard II for IC
Lieferant: HONEYWELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
E2030-100ML
HONCE2030-100MLEA 308.35 EUR InStock
HONCE2030-100ML
Combined Seven Anion Standard II for IC
Standards Anionische Standards Anionische Multi-Element-Standards
Certified Reference Material (CRM).
200 mg/L PO43-, 100 mg/L each of Cl-, NO2-, Br-, NO3-, SO42-, 20 mg/L F- in H₂O
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge