Contract required detection limits (CRDL) ICP and HGA elements
Lieferant: HONEYWELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
E2107-100ML
HONCE2107-100MLEA 571.49 EUR InStock
HONCE2107-100ML
Contract required detection limits (CRDL) ICP and HGA elements
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
120 mg/L Sb, 100 mg/L each of Co, V, 80 mg/L Ni, 50 mg/L Cu, 40 mg/L Zn, 30 mg/L Mn, 20 mg/L each of Ag, As, Cr, Tl, 10 mg/L each of Be, Cd, Se, 6 mg/L Pb in HNO₃ 5%
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge