Alternate Interference A for ICP
Lieferant: HONEYWELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
E2115-100ML
HONCE2115-100MLEA 626.92 EUR InStock
HONCE2115-100ML
Alternate Interference A for ICP
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
1000 mg/L each of Cr, Cu, Mn, Ni, Ti, V in HNO₃ 5%
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge