Trace Metals I for AA and ICP-OES
Lieferant: HONEYWELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
E2166-100ML
HONCE2166-100MLEA 276.27 EUR InStock
HONCE2166-100ML
Trace Metals I for AA and ICP-OES
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
500 mg/L Al, 250 mg/L V, 100 mg/L each of As, Be, Co, Cr, Cu, Fe, Mn, Ni, Pb, Zn, 25 mg/L each of Cd, Se, 10 mg/L Hg in HNO₃ 5%
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge