E3035-100ML
HONCE3035-100MLEA
52.12
EUR
InStock
HONCE3035-100ML
Nitrite (N) standard for ion chromatography, (CRM), Fluka™
Standards
Anionische Standards
Nitrit-Standards
Certified Reference Material (CRM)
- Traceable to SRM from NIST
Zertifizierungen: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).