Organic analytical standards, Propyl 4-hydroxybenzoate-ring-13C6 solution, 50 μg/ml in acetone, reference material, Honeywell Fluka™
Lieferant: HONEYWELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
R1348-1ML
HONCR1348-1MLEA 347.55 EUR InStock
HONCR1348-1ML
Organic analytical standards, Propyl 4-hydroxybenzoate-ring-13C6 solution, 50 μg/ml in acetone, reference material, Honeywell Fluka™
Standards Organische Standards
High quality inorganic and organic reference materials produced according to ISO 9001.
  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge