Overshoes Non woven, PP
Lieferant: KOLMI HOPEN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H10020
HOPEH10020EA 211 EUR InStock
HOPEH10020
Overshoes Non woven, PP
Einweg-Kleidung
These non woven overshoes are made up of PP with Herringbone print sole.
  • Its polypropylene material is particularly used on smooth surfaces in dry environments
  • Its chevron patterns ensure grip on dry floors
Zertifizierungen: ISO 9001 manufacturing site.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge