Empty disposable sample load cartridges, RediSep®
Lieferant: COLLECTION PREFIX HPLC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AS-69-3873-235
HPLCAS-69-3873-235EA 214 EUR InStock
HPLCAS-69-3873-235
Empty disposable sample load cartridges, RediSep®
Chromatographiesäulen
RediSep® empty disposable sample load cartridges improve the resolution of the compound and eliminate reaction byproducts when compared to liquid injection techniques.
Prepare pre-filled solid load cartridges by pipetting the dissolved sample onto the top of the cartridge. Prepare empty solid load cartridges by filling the cartridge with a slurry mixture of the dissolved sample and supporting media. For optimal benefits, remove the solvent by vacuum before placing the cartridge on the purification system.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge