Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

All-in-One Printer, OfficeJet Pro 8022e

Lieferant: COLLECTION PREFIX HPLC

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

LSPHD8022E
HPLCLSPHD8022EEA 0 EUR InStock
HPLCLSPHD8022E
All-in-One Printer, OfficeJet Pro 8022e
Drucker Document Printers
Count on the fast print speeds up to 20 ppm, automatic two-sided printing, and reliable Wi-Fi connectivity.

  • Functions: Print, copy, scan, fax
  • Print speed: Print speed up to 20 ppm (black) and 10 ppm (colour)
  • Print quality black (best): 1200×1200 rendered dpi
  • Automatic document feeder capacity: Standard, 35 sheets
  • Customisable shortcuts on your phone
  • Hands-free, auto-fed
  • Printing just got more productive

Designed to save you time: Increase productivity with fast colour performance - plus fax and scan to USB - and help save time with shortcuts.

HP+. HP's smart printing system: HP+ , the smart printing system that thinks ahead so your printer is ready when you are, more secure and productive than ever. Choose HP+ and get 6 months of instant ink and one extra year of HP warranty with exclusive use of Original HP ink.

Enhanced security, reliable connections: Get security essentials to help maintain privacy and control, and self-healing Wi-Fi® that keeps you connected.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge