Safety needles, HENKE-JECT®
Lieferant: HENKE SASS WOLF
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
8300020446
HSWA8300020446EA 42.7 EUR InStock
HSWA8300020446
Safety needles, HENKE-JECT®
Injektionsnadeln
These safety needles are compatible with HENKE-JECT® syringes as well as all others syringes with Luer and Luer lock according to ISO 594.
  • Safety mechanism is an integrated part of the needle
  • Audible 'click' indicates safety activation
  • Single-handed activation
Zertifizierungen: Conform to EU-Guideline 2010/32/EU and TRBA 250.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge