Trichrome stain Kit (Masson) für die Histologie, Sigma-Aldrich®
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
Gefahr
M-Clarity™ Qualitätsstufe = MQ600
HT15-1KT
HT15-1KTEA 410 EUR InStock
HT15-1KT
Trichrome stain Kit (Masson) für die Histologie, Sigma-Aldrich®
Trichrome stain
This kit may be useful in study of diseases of connective tissue and muscle characterised by fibrotic and dystrophic changes. For in vitro diagnostic use.
Principle: Collagen and muscle fiber stain differentially when treated sequentially with Biebrich Scarlet-Acid Fuchsin, PTA/PMA and Aniline Blue. Cytoplasm and muscle fibers stain red whereas collagen displays blue coloration.
The kit contains 1× Biebrich Scarlet-acid Fuchsin solution (250 ml), 1× phosphomolybdic acid solution (250 ml), 1× phosphotungstic acid solution (250 ml) and 1× aniline blue solution (250 ml).
UN: 3316
ADR: 9 II
IVDLogo
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge