Breather controller for unistats
Lieferant: HUBER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9771
HUBE9771EA 0 EUR InStock
HUBE9771
Breather controller for unistats
Regler Utility-Controller
This breather controller enables small pressurisation of the expansion vessel.
  • Breather control limits the pressure for the pressure overlay in the expansion vessel
  • Expansion vessel in the unistats can handle a pressure of up to 0,3 bar
  • Prolonges life-time of the thermal fluid due to the reduced oxidation as well as an extended temperature range
The breather control consists of the atmospheric sealing kit and the parts built onto the mounting plate: Pressure regulator (20 – 200 mbar), needle valve, pressure relief valve, bubble counter and connection hose. The pressure regulator does have a preset pressure relief valve that will blow-off at too high a pressure.
Achtung! Bitte beachten: Not suitable for outdoor installation – Do not install in a closed room. Room must be adequately ventilated.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge