Spigot with screw caps for jerricans
Lieferant: hünersdorff GmbH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
550500
HUEN550500EA 0 EUR InStock
HUEN550500
Spigot with screw caps for jerricans
Hähne
Spigots PROFI with red DIN 60/61 screw cap is ideal for controlled emptying of liquids from industrial jerrycans.
  • Resistant to acids, alkalis and other chemicals
  • Pressure resistance up to 0,4 bar
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge