Dispenser bottle, LDPE
Lieferant: hünersdorff GmbH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
844200
HUEN844200EA 0 EUR InStock
HUEN844200
Dispenser bottle, LDPE
Flaschen Tropfflaschen
LDPE, natural, food-safe.
  • Resistant to acids and chemicals
  • Dosing by pressure on the bottle
  • With closed nozzle and protective cap
For glue, oil, and more, oval shape.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge