Equipment table
Lieferant: HUPFER METALLWERKE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
7500204
HUPF7500204EA 1620 EUR InStock
HUPF7500204
Equipment table
Möbel Tische und Werkbänke
The instrument table is made from stainless steel. It is a solid, jointless tube structure made from square-type tube (20×20 mm), with two removable table tops formed as protection gallery (border top).
  • Electro-conductive
  • With double ball bearing the thread protection
  • Castors with 75 mm diameter
The instrument table is to provide immersion trays, sterile goods containers and baskets in operating theatres and neighbouring areas.
Zertifizierungen: Conforms with DIN 90952 packaging for sterilising goods.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge