Hyclone™ Ham's F-10 Basal Media, Cell Culture Media, Liquid, Cytiva
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SH30025.01
HYCLSH30025.01EA 43.1 EUR InStock
HYCLSH30025.01
Hyclone™ Ham's F-10 Basal Media, Cell Culture Media, Liquid, Cytiva
Zellkulturmedien
Ham's F-10 was designed for use with Chinese Hamster Ovary (CHO) cells, and has been shown to support the growth of human diploid cells, white blood cells, and primary explants from rat, rabbit, or chicken tissues. This classical media can be used for serum-free growth of some cell lines including CHO cells. Serum supplementation may or may not be necessary depending on the type of cell being cultured. Ham's F-10 contains a wide variety of components, including zinc, hypoxanthine, and thymidine and is buffered with sodium bicarbonate.
  • Contains 6,1 mM glucose
  • All liquid media products are hydrated using WFI quality water
  • 0,1 μm sterile filtered
  • Manufactured using ISO 9001:2000 certified processes
A Certificate of Analysis is included in each package with preparation instructions, intended use, and handling and performance criteria.
Bestellinformation: Liquid media are supplied in unbreakable, square PETG bottles.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge