Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

HyClone™ Calf Serum, U.S. Origin

Lieferant: GE Healthcare - Hyclone

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

SH30073.03 SH30073.02 SH30073.04 SH30073.04HI SH30073.03HI SH30073.02HI SH30073.03IR25 SH30073.02IR
HYCLSH30073.03EA 119 EUR InStock
HYCLSH30073.03 HYCLSH30073.02 HYCLSH30073.04 HYCLSH30073.04HI HYCLSH30073.03HI HYCLSH30073.02HI SH30073.03IR25 SH30073.02IR
HyClone™ Calf Serum, U.S. Origin
Seren
HyClone™ bovine calf serum can be used as an effective alternative to foetal bovine serum (FBS).

  • Filtered through sequential 100 nm pore size rated filters
  • Extensive quality, performance and analytical testing
  • Endotoxin Concentration ≤ 10 EU/mL
  • Storage Temperature is –10 °C

Origin is carefully collected, processed and filtered in U.S. abbatoirs. Ensures excellent culture results without safety concerns.

Zertifizierungen: Fully traceable and conforms to all USDA regulations.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge