Cell culture media, liquid, DMEM:F12 (1:1), HyClone™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SH30261.01 SH30023.01 SH30023.02 SH30023.FS SH30126.FS SH30271.01 SH30271.FS SH30272.02
SH30261.01EA 44.3 EUR InStock
SH30261.01 HYCLSH30023.01 HYCLSH30023.02 HYCLSH30023.FS HYCLSH30126.FS HYCLSH30271.01 HYCLSH30271.FS HYCLSH30272.02
Cell culture media, liquid, DMEM:F12 (1:1), HyClone™
Zellkulturmedien
HyClone Classical Liquid Media: DMEM F12 is manufactured using ISO 9001:2000-certified processes.
  • Contains 17,5 mM glucose
  • 0,1 µm sterile filtered
  • Manufactured using ISO 9001 certified processes
High yield cultures begin with high quality media. In order to maintain these standards, all HyClone products as well as raw material components must pass strict Quality Control testing to confirm the appropriate level of quality.
Each raw material has a specification document describing the product, the required grade or purity, the source, and the criteria for acceptance on incoming inspection. We can determine which (if any) ingredients in a medium formulation are of animal origin or, if they have come in contact with animal derived materials in manufacture.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge